missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DECR2 Full-length ORF (NP_065715.1, 1 a.a. - 292 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00026063-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16118052

  • 6845.00 SEK / 25µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MAQPPPDVEGDDCLPAYRHLFCPDLLRDKVAFITGGGSGIGFRIAEIFMRHGCHTVIASRSLPRVLTAARKLAGATGRRCLPLSMDVRAPPAVMAAVDQALKEFGRIDILINCAAGNFLCPAGALSFNAFKTVMDIDTSGTFNVSRVLYEKFFRDHGGVIVNITATLGNRGQALQVHAGSAKAAVDAMTRHLAVEWGPQNIRVNSLAPGPISGTEGLRRLGGPQASLSTKVTASPLQRLGNKTEIAHSVLYLASPLASYVTGAVLVADGGAWLTFPNGVKGLPDFASFSAKL
Specifications

Specifications

NP_065715.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
57.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PDCR/SDR17C1
DECR2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
26063
DECR2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAQPPPDVEGDDCLPAYRHLFCPDLLRDKVAFITGGGSGIGFRIAEIFMRHGCHTVIASRSLPRVLTAARKLAGATGRRCLPLSMDVRAPPAVMAAVDQALKEFGRIDILINCAAGNFLCPAGALSFNAFKTVMDIDTSGTFNVSRVLYEKFFRDHGGVIVNITATLGNRGQALQVHAGSAKAAVDAMTRHLAVEWGPQNIRVNSLAPGPISGTEGLRRLGGPQASLSTKVTASPLQRLGNKTEIAHSVLYLASPLASYVTGAVLVADGGAWLTFPNGVKGLPDFASFSAKL
RUO
DECR2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers