Learn More
Abnova™ Human DFFB Partial ORF (NP_004393, 229 a.a. - 338 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001677-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Apoptosis is a cell death process that removes toxic and/or useless cells during mammalian development. The apoptotic process is accompanied by shrinkage and fragmentation of the cells and nuclei and degradation of the chromosomal DNA into nucleosomal units. DNA fragmentation factor (DFF) is a heterodimeric protein of 40-kD (DFFB) and 45-kD (DFFA) subunits. DFFA is the substrate for caspase-3 and triggers DNA fragmentation during apoptosis. DFF becomes activated when DFFA is cleaved by caspase-3. The cleaved fragments of DFFA dissociate from DFFB, the active component of DFF. DFFB has been found to trigger both DNA fragmentation and chromatin condensation during apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene but the biological validity of these variants has not been determined. [provided by RefSeq]
Sequence: PFDMDSCLSRHSINPYSNRESRILFSTWNLDHIIEKKRTIIPTLVEAIKEQDGREVDWEYFYGLLFTSENLKLVHIVCHKKTTHKLNCDPSRIYKPQTRLKRKQPVRKRQSpecifications
NP_004393 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PFDMDSCLSRHSINPYSNRESRILFSTWNLDHIIEKKRTIIPTLVEAIKEQDGREVDWEYFYGLLFTSENLKLVHIVCHKKTTHKLNCDPSRIYKPQTRLKRKQPVRKRQ | |
RUO | |
DFFB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1677 | |
DFFB (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAD/CPAN/DFF-40/DFF2/DFF40 | |
DFFB | |
Recombinant | |
wheat germ expression system |