missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DGCR14 Full-length ORF (AAH37829.1, 1 a.a. - 395 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH37829.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8220 |
Molecular Weight (g/mol) | 67kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16192312
|
Abnova™
H00008220-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16182312
|
Abnova™
H00008220-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is located within the minimal DGS critical region (MDGCR) thought to contain the gene(s) responsible for a group of developmental disorders. These disorders include DiGeorge syndrome, velocardiofacial syndrome, conotruncal anomaly face syndrome, and some familial or sporadic conotruncal cardiac defects which have been associated with microdeletion of 22q11.2. The encoded protein may be a component of C complex spliceosomes, and the orthologous protein in the mouse localizes to the nucleus. [provided by RefSeq]
Sequence: MLGACFPNLSSGDVMVFGPGGLGQPVLHPVLVLGHHQQPVLGFGSKLVVGPVVGPQACLGVQFALSAVLALPLPLEGGRRRGFGLGGLQPRVAEDLIDVEPLADVGLQHAVDEVLALAGQVLGAREVHTVLLLDTQHLPDVGVVIGHGAADHDVQDHAQAPDVIHLGLVGDALQHLGGCICCRPAEGLAEDDAPIAVPQAALGEAKVRQLDVEVLVEEKVLALEVPVDDVQVVAVLDGGGELSEHLACHVLMQGSLALDELEKRLAPLPSTSPRHVGQQALSSFTLPEASGWAALGWASTQGWDTQHCLRVTSANELIKPRSGGGDWAEGEQGLWNRGSQAGLAGAPTPRPLEGSLSPGQDSGGPAAAALVSLSQVKLEGRRPTSSLFASPGCGDSpecifications
AAH37829.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLGACFPNLSSGDVMVFGPGGLGQPVLHPVLVLGHHQQPVLGFGSKLVVGPVVGPQACLGVQFALSAVLALPLPLEGGRRRGFGLGGLQPRVAEDLIDVEPLADVGLQHAVDEVLALAGQVLGAREVHTVLLLDTQHLPDVGVVIGHGAADHDVQDHAQAPDVIHLGLVGDALQHLGGCICCRPAEGLAEDDAPIAVPQAALGEAKVRQLDVEVLVEEKVLALEVPVDDVQVVAVLDGGGELSEHLACHVLMQGSLALDELEKRLAPLPSTSPRHVGQQALSSFTLPEASGWAALGWASTQGWDTQHCLRVTSANELIKPRSGGGDWAEGEQGLWNRGSQAGLAGAPTPRPLEGSLSPGQDSGGPAAAALVSLSQVKLEGRRPTSSLFASPGCGD | |
RUO | |
DGCR14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8220 | |
DGCR14 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DGCR13/DGS-H/DGS-I/DGSH/DGSI/ES2/Es2el | |
DGCR14 | |
Recombinant | |
wheat germ expression system |