Learn More
Abnova™ Human DGKE Partial ORF (NP_003638, 141 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008526-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Diacylglycerol kinases are thought to be involved mainly in the regeneration of phosphatidylinositol (PI) from diacylglycerol in the PI-cycle during cell signal transduction. When expressed in mammalian cells, DGK-epsilon shows specificity for arachidonyl-containing diacylglycerol. DGK-epsilon is expressed predominantly in testis. [provided by RefSeq]
Sequence: CKQQCGCQPKLCDYRCIWCQKTVHDECMKNSLKNEKCDFGEFKNLIIPPSYLTSINQMRKDKKTDYEVLASKLGKQWTPLIILANSRSGTNMGEGLLGEFSpecifications
NP_003638 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CKQQCGCQPKLCDYRCIWCQKTVHDECMKNSLKNEKCDFGEFKNLIIPPSYLTSINQMRKDKKTDYEVLASKLGKQWTPLIILANSRSGTNMGEGLLGEF | |
RUO | |
DGKE | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8526 | |
DGKE (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DAGK6/DGK | |
DGKE | |
Recombinant | |
wheat germ expression system |