Learn More
Abnova™ Human DHX32 Full-length ORF (AAH02473.3, 1 a.a. - 743 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH02473.3 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55760 |
Molecular Weight (g/mol) | 110.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16131773
|
Abnova™
H00055760-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16141773
|
Abnova™
H00055760-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a member of this family. The function of this member has not been determined. Alternative splicing of this gene generates 2 transcript variants, but the full length nature of one of the variants has not been defined. [provided by RefSeq]
Sequence: MEEEGLECPNSSSEKRYFPESLDSSDGDEEEVLACEDLELNPFDGLPYSSRYYKLLKEREDLPIWKEKYSFMENLLQNQIVIVSGDAKCGKSAQVPQWCAEYCLSIHYQHGGVICTQVHKQTVVQLALRVADEMDVNIGHEVGYVIPFENCCTNETILRYCTDDMLQREMMSNPFLGSYGVIILDDIHERSIATDVLLGLLKDVLLARPELKLIINSSPHLISKLNSYYGNVPVIEVKNKHPVEVVYLSEAQKDSFESILRLIFEIHHSGEKGDIVVFLACEQDIEKVCETVYQGSNLNPDLGELVVVPLYPKEKCSLFKPLDETEKRCQVYQRRVVLTTSSGEFLIWSNSVRFVIDVGVERRKVYNPRIRANSLVMQPISQSQAEIRKQILGSSSSGKFFCLYTEEFASKDMTPLKPAEMQEANLTSMVLFMKRIDIAGLGHCDFMNRPAPESLMQALEDLDYLAALDNDGNLSEFGIIMSEFPLDPQLSKSILASCEFDCVDEVLTIAAMVTAPNCFSHVPHGAEEAALTCWKTFLHPEGDHFTLISIYKAYQDTTLNSSSEYCVEKWCRDYFLNCSALRMADVIRAELLEIIKRIELPYAEPAFGSKENTLNIKKALLSGYFMQIARDVDGSGNYLMLTHKQVAQLHPLSGYSITKKMPEWVLFHKFSISENNYIRITSEISPELFMQLVPQYYFSNLPPSESKDILQQVVDHLSPVSTMNKEQQMCETCPETEQRCTLQSpecifications
AAH02473.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEEEGLECPNSSSEKRYFPESLDSSDGDEEEVLACEDLELNPFDGLPYSSRYYKLLKEREDLPIWKEKYSFMENLLQNQIVIVSGDAKCGKSAQVPQWCAEYCLSIHYQHGGVICTQVHKQTVVQLALRVADEMDVNIGHEVGYVIPFENCCTNETILRYCTDDMLQREMMSNPFLGSYGVIILDDIHERSIATDVLLGLLKDVLLARPELKLIINSSPHLISKLNSYYGNVPVIEVKNKHPVEVVYLSEAQKDSFESILRLIFEIHHSGEKGDIVVFLACEQDIEKVCETVYQGSNLNPDLGELVVVPLYPKEKCSLFKPLDETEKRCQVYQRRVVLTTSSGEFLIWSNSVRFVIDVGVERRKVYNPRIRANSLVMQPISQSQAEIRKQILGSSSSGKFFCLYTEEFASKDMTPLKPAEMQEANLTSMVLFMKRIDIAGLGHCDFMNRPAPESLMQALEDLDYLAALDNDGNLSEFGIIMSEFPLDPQLSKSILASCEFDCVDEVLTIAAMVTAPNCFSHVPHGAEEAALTCWKTFLHPEGDHFTLISIYKAYQDTTLNSSSEYCVEKWCRDYFLNCSALRMADVIRAELLEIIKRIELPYAEPAFGSKENTLNIKKALLSGYFMQIARDVDGSGNYLMLTHKQVAQLHPLSGYSITKKMPEWVLFHKFSISENNYIRITSEISPELFMQLVPQYYFSNLPPSESKDILQQVVDHLSPVSTMNKEQQMCETCPETEQRCTLQ | |
RUO | |
DHX32 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55760 | |
DHX32 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DDX32/DHLP1/FLJ10694/FLJ10889 | |
DHX32 | |
Recombinant | |
wheat germ expression system |