Learn More
Abnova™ Human DMBX1 Partial ORF (NP_671725, 1 a.a. - 88 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00127343-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MQHYGVNGYSLHAMNSLSAMYNLHQQAAQQAQHAPDYRPSVHALTLAERLAGCTFQDIILEARYGSQHRKQRRSRTAFTAQQLEALEKSpecifications
NP_671725 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQHYGVNGYSLHAMNSLSAMYNLHQQAAQQAQHAPDYRPSVHALTLAERLAGCTFQDIILEARYGSQHRKQRRSRTAFTAQQLEALEK | |
RUO | |
DMBX1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
127343 | |
DMBX1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MBX/OTX3/PAXB | |
DMBX1 | |
Recombinant | |
wheat germ expression system |