Learn More
Abnova™ Human DMTF1 Partial ORF (NP_066968, 661 a.a. - 760 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009988-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a transcription factor that contains a cyclin D-binding domain, three central Myb-like repeats, and two flanking acidic transactivation domains at the N- and C-termini. The encoded protein is induced by the oncogenic Ras signaling pathway and functions as a tumor suppressor by activating the transcription of ARF and thus the ARF-p53 pathway to arrest cell growth or induce apoptosis. It also activates the transcription of aminopeptidase N and may play a role in hematopoietic cell differentiation. The transcriptional activity of this protein is regulated by binding of D-cyclins. This gene is hemizygously deleted in approximately 40% of human non-small-cell lung cancer and is a potential prognostic and gene-therapy target for non-small-cell lung cancer. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: KQEESPSDLASAYVTEGLESPTIEEQVDQTIDDETILIVPSPHGFIQASDVIDTESVLPLTTLTDPILQHHQEESNIIGSSLGSPVSEDSKDVEDLVNCHSpecifications
NP_066968 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KQEESPSDLASAYVTEGLESPTIEEQVDQTIDDETILIVPSPHGFIQASDVIDTESVLPLTTLTDPILQHHQEESNIIGSSLGSPVSEDSKDVEDLVNCH | |
RUO | |
DMTF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9988 | |
DMTF1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DMP1/DMTF/FLJ25188/FLJ41265/FLJ76054/hDMP1 | |
DMTF1 | |
Recombinant | |
wheat germ expression system |