missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DOHH Full-length ORF (NP_112594.1, 1 a.a. - 302 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_112594.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 83475 |
Molecular Weight (g/mol) | 59.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16135353
|
Abnova™
H00083475-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16125353
|
Abnova™
H00083475-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a metalloenzyme that catalyzes the last step in the conversion of lysine to the unique amino acid hypusine in eukaryotic initiation factor 5A. The encoded protein hydroxylates deoxyhypusine to form hypusine in the mature eukaryotic initiation factor 5A protein. Alternative splicing results in multiple transcript variants
Sequence: MVTEQEVDAIGQTLVDPKQPLQARFRALFTLRGLGGPGAIAWISQAFDDDSALLKHELAYCLGQMQDARAIPMLVDVLQDTRQEPMVRHEAGEALGAIGDPEVLEILKQYSSDPVIEVAETCQLAVRRLEWLQQHGGEPAAGPYLSVDPAPPAEERDVGRLREALLDESRPLFERYRAMFALRNAGGEEAALALAEGLHCGSALFRHEVGYVLGQLQHEAAVPQLAAALARCTENPMVRHECAEALGAIARPACLAALQAHADDPERVVRESCEVALDMYEHETGRAFQYADGLEQLRGAPSSpecifications
NP_112594.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MVTEQEVDAIGQTLVDPKQPLQARFRALFTLRGLGGPGAIAWISQAFDDDSALLKHELAYCLGQMQDARAIPMLVDVLQDTRQEPMVRHEAGEALGAIGDPEVLEILKQYSSDPVIEVAETCQLAVRRLEWLQQHGGEPAAGPYLSVDPAPPAEERDVGRLREALLDESRPLFERYRAMFALRNAGGEEAALALAEGLHCGSALFRHEVGYVLGQLQHEAAVPQLAAALARCTENPMVRHECAEALGAIARPACLAALQAHADDPERVVRESCEVALDMYEHETGRAFQYADGLEQLRGAPS | |
RUO | |
DOHH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
83475 | |
DOHH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HLRC1/MGC4293 | |
DOHH | |
Recombinant | |
wheat germ expression system |