Learn More
Abnova™ Human DOHH Full-length ORF (NP_112594.1, 1 a.a. - 302 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00083475-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a metalloenzyme that catalyzes the last step in the conversion of lysine to the unique amino acid hypusine in eukaryotic initiation factor 5A. The encoded protein hydroxylates deoxyhypusine to form hypusine in the mature eukaryotic initiation factor 5A protein. Alternative splicing results in multiple transcript variants
Sequence: MVTEQEVDAIGQTLVDPKQPLQARFRALFTLRGLGGPGAIAWISQAFDDDSALLKHELAYCLGQMQDARAIPMLVDVLQDTRQEPMVRHEAGEALGAIGDPEVLEILKQYSSDPVIEVAETCQLAVRRLEWLQQHGGEPAAGPYLSVDPAPPAEERDVGRLREALLDESRPLFERYRAMFALRNAGGEEAALALAEGLHCGSALFRHEVGYVLGQLQHEAAVPQLAAALARCTENPMVRHECAEALGAIARPACLAALQAHADDPERVVRESCEVALDMYEHETGRAFQYADGLEQLRGAPSSpecifications
NP_112594.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HLRC1/MGC4293 | |
DOHH | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
83475 | |
DOHH (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVTEQEVDAIGQTLVDPKQPLQARFRALFTLRGLGGPGAIAWISQAFDDDSALLKHELAYCLGQMQDARAIPMLVDVLQDTRQEPMVRHEAGEALGAIGDPEVLEILKQYSSDPVIEVAETCQLAVRRLEWLQQHGGEPAAGPYLSVDPAPPAEERDVGRLREALLDESRPLFERYRAMFALRNAGGEEAALALAEGLHCGSALFRHEVGYVLGQLQHEAAVPQLAAALARCTENPMVRHECAEALGAIARPACLAALQAHADDPERVVRESCEVALDMYEHETGRAFQYADGLEQLRGAPS | |
RUO | |
DOHH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |