missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DPH2 Full-length ORF (AAH16956, 1 a.a. - 489 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH16956 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1802 |
Molecular Weight (g/mol) | 79.53kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16144161
|
Abnova™
H00001802-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16134161
|
Abnova™
H00001802-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene is one of two human genes similar to the yeast gene dph2. The yeast gene was identified by its ability to complement a diphthamide mutant strain, and thus probably functions in diphthamide biosynthesis. Diphthamide is a post-translationally modified histidine residue present in elongation factor 2 (EF2) that is the target of diphtheria toxin ADP-ribosylation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MESMFSSPAEAALQRETGVPGLLTPLPDLDGVYELERVAGFVRDLGCERVALQFPDQLLGDAVAVAARLEETTGSKMFILGDTAYGSCCVDVLGAEQAGAQALIHFGPACLSPPARPLPVAFVLRQRSVALELCVKAFEAQNPDPKAPVVLLSEPACAHALEALATLLRPRYLDLLVSSPAFPQPVGSLSPEPMPLERFGRRFPLAPGRRLEEYGAFYVGGSKASPDPDLDPDLSRLLLGWAPGQPFSSCCPDTGKTQDEGARAGRLRARRRYLVERARDARVVGLLAGTLGVAQHREALAHLRNLTQAAGKRSYVLALGRPTPAKLANFPEVDVFVLLACPLGALAPQLSGSFFQPILAPCELEAACNPAWPPPGLAPHLTHYADLLPGSPFHVALPPPESELWETPDVSLITGDLRPPPAWKSSNDHGSLALTPRPQLELAESSPAASFLSSRSWQGLEPRLGQTPVTEAVSGRRGIAIAYEDEGSGSpecifications
AAH16956 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.53kDa | |
Glutathione Sepharose 4 Fast Flow | |
MESMFSSPAEAALQRETGVPGLLTPLPDLDGVYELERVAGFVRDLGCERVALQFPDQLLGDAVAVAARLEETTGSKMFILGDTAYGSCCVDVLGAEQAGAQALIHFGPACLSPPARPLPVAFVLRQRSVALELCVKAFEAQNPDPKAPVVLLSEPACAHALEALATLLRPRYLDLLVSSPAFPQPVGSLSPEPMPLERFGRRFPLAPGRRLEEYGAFYVGGSKASPDPDLDPDLSRLLLGWAPGQPFSSCCPDTGKTQDEGARAGRLRARRRYLVERARDARVVGLLAGTLGVAQHREALAHLRNLTQAAGKRSYVLALGRPTPAKLANFPEVDVFVLLACPLGALAPQLSGSFFQPILAPCELEAACNPAWPPPGLAPHLTHYADLLPGSPFHVALPPPESELWETPDVSLITGDLRPPPAWKSSNDHGSLALTPRPQLELAESSPAASFLSSRSWQGLEPRLGQTPVTEAVSGRRGIAIAYEDEGSG | |
RUO | |
DPH2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1802 | |
DPH2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DPH2L2 | |
DPH2 | |
Recombinant | |
wheat germ expression system |