missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human DPP6 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2360
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
DPP6 Polyclonal specifically detects DPP6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-ready.
Specifikationer
| DPP6 | |
| Polyclonal | |
| Unconjugated | |
| AAA35761 | |
| DPP6 | |
| Mouse myeloma cell line NS0-derived recombinant human DPP6 Thr56-Asp803 (Glu458Val) Accession # AAA35761 | |
| 100 μg | |
| Primary | |
| Detects human DPP6 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, CyTOF-ready | |
| dipeptidyl aminopeptidase IV-related protein, dipeptidyl aminopeptidase-like protein 6, Dipeptidyl aminopeptidase-related protein, Dipeptidyl peptidase 6, Dipeptidyl peptidase IV-like protein, dipeptidyl peptidase IV-related protein, Dipeptidyl peptidase VI, dipeptidylpeptidase 6, dipeptidyl-peptidase 6, dipeptidylpeptidase VI, DPP VI, DPPX, DPPXFLJ55680, MGC46605, VF2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1804 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering