Learn More
Abnova™ Human DPYD Partial ORF (NP_000101, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001806-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a pyrimidine catabolic enzyme and the initial and rate-limiting factor in the pathway of uracil and thymidine catabolism. Mutations in this gene result in dihydropyrimidine dehydrogenase deficiency, an error in pyrimidine metabolism associated with thymine-uraciluria and an increased risk of toxicity in cancer patients receiving 5-fluorouracil chemotherapy. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MAPVLSKDSADIESILALNPRTQTHATLCSTSAKKLDKKHWKRNPDKNCFNCEKLENNFDDIKHTTLGERGALREAMRCLKCADAPCQKSCPTNLDIKSFITSIANKNYYSpecifications
NP_000101 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAPVLSKDSADIESILALNPRTQTHATLCSTSAKKLDKKHWKRNPDKNCFNCEKLENNFDDIKHTTLGERGALREAMRCLKCADAPCQKSCPTNLDIKSFITSIANKNYY | |
RUO | |
DPYD | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1806 | |
DPYD (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DHP/DHPDHASE/DPD/MGC132008/MGC70799 | |
DPYD | |
Recombinant | |
wheat germ expression system |