missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DRF1 Partial ORF (NP_663696, 201 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_663696 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 80174 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16150197
|
Abnova™
H00080174-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16140197
|
Abnova™
H00080174-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a regulator of the CDC7-like 1 protein, a serine-threonine kinase which links cell cycle regulation to genome duplication. This protein localizes to the nucleus and its expression is cell cycle-regulated. Alternative splicing of this gene results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full length sequences have not been determined. [provided by RefSeq]
Sequence: VKKQQPKKPEGTCPAAESRTRKVARLKAPFLKIEDESRKFRPFHHQFKSFPEISFLGPKDASPFEAPTTLGSMHHTRESKDGEPSPRSAAHTMPRRKKGSpecifications
NP_663696 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASKL1/DRF1/FLJ13087/MGC15009/ZDBF1B/chifb | |
DBF4B | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
80174 | |
DRF1 (Human) Recombinant Protein (Q01) | |
VKKQQPKKPEGTCPAAESRTRKVARLKAPFLKIEDESRKFRPFHHQFKSFPEISFLGPKDASPFEAPTTLGSMHHTRESKDGEPSPRSAAHTMPRRKKG | |
RUO | |
DBF4B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |