Learn More
Abnova™ Human DSC3 Partial ORF (NP_001932, 585 a.a. - 684 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_001932 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1825 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124201
|
Abnova™
H00001825-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16114201
|
Abnova™
H00001825-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a calcium-dependent glycoprotein that is a member of the desmocollin subfamily of the cadherin superfamily. These desmosomal family members, along with the desmogleins, are found primarily in epithelial cells where they constitute the adhesive proteins of the desmosome cell-cell junction and are required for cell adhesion and desmosome formation. The desmosomal family members are arranged in two clusters on chromosome 18, occupying less than 650 kb combined. Alternative splicing results in two transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: CKPKMGYTDILAVDPDEPVHGAPFYFSLPNTSPEISRLWSLTKVNDTAARLSYQKNAGFQEYTIPITVKDRAGQAATKLLRVNLCECTHPTQCRATSRSTSpecifications
NP_001932 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDHF3/DSC/DSC1/DSC2/DSC4/HT-CP | |
DSC3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1825 | |
DSC3 (Human) Recombinant Protein (Q01) | |
CKPKMGYTDILAVDPDEPVHGAPFYFSLPNTSPEISRLWSLTKVNDTAARLSYQKNAGFQEYTIPITVKDRAGQAATKLLRVNLCECTHPTQCRATSRST | |
RUO | |
DSC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |