Learn More
Abnova™ Human DTD1 Full-length ORF (NP_543010.3, 1 a.a. - 209 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00092675-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is similar in sequence to histidyl-tRNA synthetase, which hydrolyzes D-tyrosyl-tRNA(Tyr) into D-tyrosine and free tRNA(Tyr). The encoded protein is found in the cytoplasm. [provided by RefSeq]
Sequence: MKAVVQRVTRASVTVGGEQISAIGRGICVLLGISLEDTQKELEHMVRKILNLRVFEDESGKHWSKSVMDKQYEILCVSQFTLQCVLKGNKPDFHLAMPTEQAEGFYNSFLEQLRKTYRPELIKDGKFGAYMQVHIQNDGPVTIELESPAPGTATSDPKQLSKLEKQQQRKEKTRAKGPSESSKERNTPRKEDRSASSGAEGDVSSEREPSpecifications
NP_543010.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C20orf88/DUEB/HARS2/MGC119131/MGC41905/bA379J5.3/bA555E18.1/pqn-68 | |
DTD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
92675 | |
DTD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKAVVQRVTRASVTVGGEQISAIGRGICVLLGISLEDTQKELEHMVRKILNLRVFEDESGKHWSKSVMDKQYEILCVSQFTLQCVLKGNKPDFHLAMPTEQAEGFYNSFLEQLRKTYRPELIKDGKFGAYMQVHIQNDGPVTIELESPAPGTATSDPKQLSKLEKQQQRKEKTRAKGPSESSKERNTPRKEDRSASSGAEGDVSSEREP | |
RUO | |
DTD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |