missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DUSP8 Partial ORF (NP_004411, 2 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001850-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16114261

  • 4515.00 SEK / 10µg
Estimated Shipment: 19-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the dual specificity protein phosphatase subfamily. These phosphatases inactivate their target kinases by dephosphorylating both the phosphoserine/threonine and phosphotyrosine residues. They negatively regulate members of the mitogen-activated protein (MAP) kinase superfamily (MAPK/ERK, SAPK/JNK, p38), which is associated with cellular proliferation and differentiation. Different members of the family of dual specificity phosphatases show distinct substrate specificities for various MAP kinases, different tissue distribution and subcellular localization, and different modes of inducibility of their expression by extracellular stimuli. This gene product inactivates SAPK/JNK and p38, is expressed predominantly in the adult brain, heart, and skeletal muscle, is localized in the cytoplasm, and is induced by nerve growth factor and insulin. An intronless pseudogene for DUSP8 is present on chromosome 10q11.2. [provided by RefSeq]

Sequence: AGDRLPRKVMDAKKLASLLRGGPGGPLVIDSRSFVEYNSWHVLSSVNICCSKLVKRRLQQGKVTIAELIQPAARSQVEATEPQDVVVYDQSTRDASVLAADSFLSILL
Specifications

Specifications

NP_004411
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AGDRLPRKVMDAKKLASLLRGGPGGPLVIDSRSFVEYNSWHVLSSVNICCSKLVKRRLQQGKVTIAELIQPAARSQVEATEPQDVVVYDQSTRDASVLAADSFLSILL
RUO
DUSP8
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1850
DUSP8 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C11orf81/FLJ42476/FLJ42958/HB5/HVH-5/HVH8
DUSP8
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers