Learn More
Abnova™ Human E2F7 Partial ORF (NP_976328.1, 801 a.a. - 910 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00144455-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
E2F transcription factors, such as E2F7, play an essential role in the regulation of cell cycle progression (Di Stefano et al., 2003 [PubMed 14633988]).[supplied by OMIM]
Sequence: VVNPKSSTLPSADPQLQSQPSLNLSPVMSRSHSVVQQPESPVYVGHPVSVVKLHQSPVPVTPKSIQRTHRETFFKTPGSLGDPVLKRRERNQSRNTSSAQRRLEIPSGGASpecifications
NP_976328.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VVNPKSSTLPSADPQLQSQPSLNLSPVMSRSHSVVQQPESPVYVGHPVSVVKLHQSPVPVTPKSIQRTHRETFFKTPGSLGDPVLKRRERNQSRNTSSAQRRLEIPSGGA | |
RUO | |
E2F7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
144455 | |
E2F7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12981 | |
E2F7 | |
Recombinant | |
wheat germ expression system |