Learn More
Abnova™ Human EDG1 Partial ORF (AAH18650, 1 a.a. - 47 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001901-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is structurally similar to G protein-coupled receptors and is highly expressed in endothelial cells. It binds the ligand sphingosine-1-phosphate with high affinity and high specificity, and suggested to be involved in the processes that regulate the differentiation of endothelial cells. Activation of this receptor induces cell-cell adhesion. [provided by RefSeq]
Sequence: MGPTSVPLVKAHRSSVSDYVNYDIIVRHYNYTGKLNISADKENSIKLSpecifications
AAH18650 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
30.91kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGPTSVPLVKAHRSSVSDYVNYDIIVRHYNYTGKLNISADKENSIKL | |
RUO | |
S1PR1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1901 | |
EDG1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CHEDG1/D1S3362/ECGF1/EDG-1/EDG1/FLJ58121/S1P1 | |
S1PR1 | |
Recombinant | |
wheat germ expression system |