missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EEF1D Full-length ORF (NP_001951.2, 1 a.a. - 281 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001936-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137307

  • 4515.00 SEK / 10µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene. [provided by RefSeq]

Sequence: MATNFLAHEKIWFDKFKYDDAERRFYEQMNGPVAGASRQENGASVILRDIARARENIQKSLAGSSGPGASSGTSGDHGELVVRIASLEVENQSLRGVVQELQQAISKLEARLNVLEKSSPGHRATAPQTQHVSPMRQVEPPAKKPATPAEDDEDDDIDLFGSDNEEEDKEAAQLREERLRQYAEKKAKKPALVAKSSILLDVKPWDDETDMAQLEACVRSIQLDGLVWGASKLVPVGYGIRKLQIQCVVEDDKVGTDLLEEEITKFEEHVQSVDIAAFNKI
Specifications

Specifications

NP_001951.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
57.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EF-1D/EF1D/FLJ20897/FP1047
EEF1D
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
1936
EEF1D (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATNFLAHEKIWFDKFKYDDAERRFYEQMNGPVAGASRQENGASVILRDIARARENIQKSLAGSSGPGASSGTSGDHGELVVRIASLEVENQSLRGVVQELQQAISKLEARLNVLEKSSPGHRATAPQTQHVSPMRQVEPPAKKPATPAEDDEDDDIDLFGSDNEEEDKEAAQLREERLRQYAEKKAKKPALVAKSSILLDVKPWDDETDMAQLEACVRSIQLDGLVWGASKLVPVGYGIRKLQIQCVVEDDKVGTDLLEEEITKFEEHVQSVDIAAFNKI
RUO
EEF1D
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers