missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EEF1D Partial ORF (NP_115754, 1 a.a. - 91 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_115754 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1936 |
Molecular Weight (g/mol) | 35.75kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137484
|
Abnova™
H00001936-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16127484
|
Abnova™
H00001936-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene. [provided by RefSeq]
Sequence: MRSGKASCTLETVWEDKHKYEEAERRFYEHEATQAAASAQQLPAEGPAMNGPGQDDPEDADEAEAPDGGSRRDPRKSQDSRKPLQKKRKRSSpecifications
NP_115754 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EF-1D/EF1D/FLJ20897/FP1047 | |
EEF1D | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1936 | |
EEF1D (Human) Recombinant Protein (Q01) | |
MRSGKASCTLETVWEDKHKYEEAERRFYEHEATQAAASAQQLPAEGPAMNGPGQDDPEDADEAEAPDGGSRRDPRKSQDSRKPLQKKRKRS | |
RUO | |
EEF1D | |
Wheat Germ (in vitro) | |
GST | |
Liquid |