Learn More
Abnova™ Human EFNA2 Full-length ORF (AAI48728.1, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAI48728.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1943 |
Molecular Weight (g/mol) | 50.38kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16134391
|
Abnova™
H00001943-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16124391
|
Abnova™
H00001943-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the ephrin family. The protein is composed of a signal sequence, a receptor-binding region, a spacer region, and a hydrophobic region. The EPH and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. Posttranslational modifications determine whether this protein localizes to the nucleus or the cytoplasm. [provided by RefSeq]
Sequence: MAPAQRPLLPLLLLLLPLPPPPFARAEDAARANSDRYAVYWNRSNPRFHAGAGDDGGGYTVEVSINDYLDIYCPHYGAPLPPAERMEHYVLYMVNGEGHASCDHRQRGFKRWECNRPAAPGGPLKFSEKFQLFTPFSLGFEFRPGHEYYYISATPPNAVDRPCLRLKVYVRPTNETLYEAPEPIFTSNNSCSSPGGCRLFLSTIPVLWTLLGSSpecifications
AAI48728.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.38kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAPAQRPLLPLLLLLLPLPPPPFARAEDAARANSDRYAVYWNRSNPRFHAGAGDDGGGYTVEVSINDYLDIYCPHYGAPLPPAERMEHYVLYMVNGEGHASCDHRQRGFKRWECNRPAAPGGPLKFSEKFQLFTPFSLGFEFRPGHEYYYISATPPNAVDRPCLRLKVYVRPTNETLYEAPEPIFTSNNSCSSPGGCRLFLSTIPVLWTLLGS | |
RUO | |
EFNA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1943 | |
EFNA2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ELF-1/EPLG6/HEK7-L/LERK6 | |
EFNA2 | |
Recombinant | |
wheat germ expression system |