Learn More
Abnova™ Human EFNB2 Full-length ORF (NP_004084.1, 1 a.a. - 333 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001948-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ephrin (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. This gene encodes an EFNB class ephrin which binds to the EPHB4 and EPHA3 receptors. [provided by RefSeq]
Sequence: MAVRRDSVWKYCWGVLMVLCRTAISKSIVLEPIYWNSSNSKFLPGQGLVLYPQIGDKLDIICPKVDSKTVGQYEYYKVYMVDKDQADRCTIKKENTPLLNCAKPDQDIKFTIKFQEFSPNLWGLEFQKNKDYYIISTSNGSLEGLDNQEGGVCQTRAMKILMKVGQDASSAGSTRNKDPTRRPELEAGTNGRSSTTSPFVKPNPGSSTDGNSAGHSGNNILGSEVALFAGIASGCIIFIVIIITLVVLLLKYRRRHRKHSPQHTTTLSLSTLATPKRSGNNNGSEPSDIIIPLRTADSVFCPHYEKVSGDYGHPVYIVQEMPPQSPANIYYKVSpecifications
NP_004084.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EPLG5/HTKL/Htk-L/LERK5/MGC126226/MGC126227/MGC126228 | |
EFNB2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
1948 | |
EFNB2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAVRRDSVWKYCWGVLMVLCRTAISKSIVLEPIYWNSSNSKFLPGQGLVLYPQIGDKLDIICPKVDSKTVGQYEYYKVYMVDKDQADRCTIKKENTPLLNCAKPDQDIKFTIKFQEFSPNLWGLEFQKNKDYYIISTSNGSLEGLDNQEGGVCQTRAMKILMKVGQDASSAGSTRNKDPTRRPELEAGTNGRSSTTSPFVKPNPGSSTDGNSAGHSGNNILGSEVALFAGIASGCIIFIVIIITLVVLLLKYRRRHRKHSPQHTTTLSLSTLATPKRSGNNNGSEPSDIIIPLRTADSVFCPHYEKVSGDYGHPVYIVQEMPPQSPANIYYKV | |
RUO | |
EFNB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |