missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EIF1AY Partial ORF (NP_004672.2, 22 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004672.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9086 |
Molecular Weight (g/mol) | 33.88kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035125
|
Abnova™
H00009086-Q03.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16025125
|
Abnova™
H00009086-Q03.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein similar to eukaryotic translation initiation factor 1A (EIF1A). EIF1A is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA. [provided by RefSeq]
Sequence: EKRELVFKEDGQEYAQVIKMLGNGRLEALCFDGVKRLCHIRGKLRKKVWINTSDIILVGLRDYQDNKADVILKYSpecifications
NP_004672.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.88kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EIF1AY | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9086 | |
EIF1AY (Human) Recombinant Protein (Q03) | |
EKRELVFKEDGQEYAQVIKMLGNGRLEALCFDGVKRLCHIRGKLRKKVWINTSDIILVGLRDYQDNKADVILKY | |
RUO | |
EIF1AY | |
Recombinant | |
wheat germ expression system |