Learn More
Abnova™ Human EIF4EBP3 Full-length ORF (AAH10881, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008637-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the EIF4EBP family which derives it name from proteins that bind to eukaryotic initiation factor 4E and that prevent its assembly into EIF4F. Co-transcription of this gene and the neighboring upstream gene (MASK) generates a transcript (MASK-BP3) which encodes a fusion protein comprised of the MASK protein sequence for the majority of the protein and a different C-terminus due to an alternate reading frame for the EIF4EBP3 segments. [provided by RefSeq]
Sequence: MSTSTSCPIPGGRDQLPDCYSTTPGGTLYATTPGGTRIIYDRKFLLECKNSPIARTPPCCLPQIPGVTTPPTAPLSKLEELKEQETEEEIPDDAQFEMDISpecifications
AAH10881 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
4E-BP3 | |
EIF4EBP3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8637 | |
EIF4EBP3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSTSTSCPIPGGRDQLPDCYSTTPGGTLYATTPGGTRIIYDRKFLLECKNSPIARTPPCCLPQIPGVTTPPTAPLSKLEELKEQETEEEIPDDAQFEMDI | |
RUO | |
EIF4EBP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |