Learn More
Abnova™ Human EMB Full-length ORF (NP_940851.1, 1 a.a. - 327 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00133418-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a transmembrane glycoprotein that is a member of the immunoglobulin superfamily. The encoded protein may be involved in cell growth and development by mediating interactions between the cell and extracellular matrix. A pseudogene of this gene is found on chromosome 1. [provided by RefSeq]
Sequence: MRALPGLLEARARTPRLLLLQCLLAAARPSSADGSAPDSPFTSPPLREEIMANNFSLESHNISLTEHSSMPVEKNITLERPSNVNLTCQFTTSGDLNAVNVTWKKDGEQLENNYLVSATGSTLYTQYRFTIINSKQMGSYSCFFREEKEQRGTFNFKVPELHGKNKPLISYVGDSTVLTCKCQNCFPLNWTWYSSNGSVKVPVGVQMNKYVINGTYANETKLKITQLLEEDGESYWCRALFQLGESEEHIELVVLSYLVPLKPFLVIVAEVILLVATILLCEKYTQKKKKHSDEGKEFEQIEQLKSDDSNGIENNVPRHRKNESLGQSpecifications
NP_940851.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC71745 | |
EMB | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
133418 | |
EMB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRALPGLLEARARTPRLLLLQCLLAAARPSSADGSAPDSPFTSPPLREEIMANNFSLESHNISLTEHSSMPVEKNITLERPSNVNLTCQFTTSGDLNAVNVTWKKDGEQLENNYLVSATGSTLYTQYRFTIINSKQMGSYSCFFREEKEQRGTFNFKVPELHGKNKPLISYVGDSTVLTCKCQNCFPLNWTWYSSNGSVKVPVGVQMNKYVINGTYANETKLKITQLLEEDGESYWCRALFQLGESEEHIELVVLSYLVPLKPFLVIVAEVILLVATILLCEKYTQKKKKHSDEGKEFEQIEQLKSDDSNGIENNVPRHRKNESLGQ | |
RUO | |
EMB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |