missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EMP3 Partial ORF (AAH09718.1, 26 a.a. - 75 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002014-Q02.25ug
Additional Details : Weight : 0.02000kg
Description
Sequence: KSWWTLPGKESLNLWYDCTWNNDTKTWACSNVSENGWLKAVQVLMVLSLISpecifications
AAH09718.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
31.13kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KSWWTLPGKESLNLWYDCTWNNDTKTWACSNVSENGWLKAVQVLMVLSLI | |
RUO | |
EMP3 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
2014 | |
EMP3 (Human) Recombinant Protein (Q02) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
YMP | |
EMP3 | |
Yes |