Learn More
Abnova™ Human ENC1 Full-length ORF (NP_003624.1, 1 a.a. - 589 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_003624.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8507 |
Molecular Weight (g/mol) | 92.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16132512
|
Abnova™
H00008507-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16122512
|
Abnova™
H00008507-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DNA damage and/or hyperproliferative signals activate wildtype p53 tumor suppressor protein (TP53; MIM 191170), inducing cell cycle arrest or apoptosis. Mutations that inactivate p53 occur in 50% of all tumors. Polyak et al. (1997) [PubMed 9305847] used serial analysis of gene expression (SAGE) to evaluate cellular mRNA levels in a colorectal cancer cell line transfected with p53. Of 7,202 transcripts identified, only 14 were expressed at levels more than 10-fold higher in p53-expressing cells than in control cells. Polyak et al. (1997) [PubMed 9305847] termed these genes 'p53-induced genes,' or PIGs, several of which were predicted to encode redox-controlling proteins. They noted that reactive oxygen species (ROS) are potent inducers of apoptosis. Flow cytometric analysis showed that p53 expression induces ROS production, which increases as apoptosis progresses under some conditions. The authors stated that the PIG10 gene, also called ENC1, encodes an actin-binding protein.[supplied by OMIM]
Sequence: MSVSVHENRKSRASSGSINIYLFHKSSYADSVLTHLNLLRQQRLFTDVLLHAGNRTFPCHRAVLAACSRYFEAMFSGGLKESQDSEVNFDNSIHPEVLELLLDYAYSSRVIINEENAESLLEAGDMLEFQDIRDACAEFLEKNLHPTNCLGMLLLSDAHQCTKLYELSWRMCLSNFQTIRKNEDFLQLPQDMVVQLLSSEELETEDERLVYESAINWISYDLKKRYCYLPELLQTVRLALLPAIYLMENVAMEELITKQRKSKEIVEEAIRCKLKILQNDGVVTSLCARPRKTGHALFLLGGQTFMCDKLYLVDQKAKEIIPKADIPSPRKEFSACAIGCKVYITGGRGSENGVSKDVWVYDTLHEEWSKAAPMLVARFGHGSAELKHCLYVVGGHTAATGCLPASPSVSLKQVEHYDPTINKWTMVAPLREGVSNAAVVSAKLKLFAFGGTSVSHDKLPKVQCYDQCENRWTVPATCPQPWRYTAAAVLGNQIFIMGGDTEFSACSAYKFNSETYQWTKVGDVTAKRMSCHAVASGNKLYVVGGYFGIQRCKTLDCYDPTLDVWNSITTVPYSLIPTAFVSTWKHLPSSpecifications
NP_003624.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
92.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSVSVHENRKSRASSGSINIYLFHKSSYADSVLTHLNLLRQQRLFTDVLLHAGNRTFPCHRAVLAACSRYFEAMFSGGLKESQDSEVNFDNSIHPEVLELLLDYAYSSRVIINEENAESLLEAGDMLEFQDIRDACAEFLEKNLHPTNCLGMLLLSDAHQCTKLYELSWRMCLSNFQTIRKNEDFLQLPQDMVVQLLSSEELETEDERLVYESAINWISYDLKKRYCYLPELLQTVRLALLPAIYLMENVAMEELITKQRKSKEIVEEAIRCKLKILQNDGVVTSLCARPRKTGHALFLLGGQTFMCDKLYLVDQKAKEIIPKADIPSPRKEFSACAIGCKVYITGGRGSENGVSKDVWVYDTLHEEWSKAAPMLVARFGHGSAELKHCLYVVGGHTAATGCLPASPSVSLKQVEHYDPTINKWTMVAPLREGVSNAAVVSAKLKLFAFGGTSVSHDKLPKVQCYDQCENRWTVPATCPQPWRYTAAAVLGNQIFIMGGDTEFSACSAYKFNSETYQWTKVGDVTAKRMSCHAVASGNKLYVVGGYFGIQRCKTLDCYDPTLDVWNSITTVPYSLIPTAFVSTWKHLPS | |
RUO | |
ENC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8507 | |
ENC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CCL28/ENC-1/FLJ39259/KLHL35/KLHL37/NRPB/PIG10/TP53I10 | |
ENC1 | |
Recombinant | |
wheat germ expression system |