missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EOMES Partial ORF (NP_005433.2, 461 a.a. - 569 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_005433.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8320 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16064995
|
Abnova™
H00008320-Q03.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054995
|
Abnova™
H00008320-Q03.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of a conserved protein family that shares a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similiar gene disrupted in mice is shown to be essential during trophoblast development and gastrulation. [provided by RefSeq]
Sequence: SHQIVPGGRYGVQSFFPEPFVNTLPQARYYNGERTVPQTNGLLSPQQSEEVANPPQRWLVTPVQQPGTNKLDISSYESEYTSSTLLPYGIKSLPLQTSHALGYYPDPTFSpecifications
NP_005433.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TBR2 | |
EOMES | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8320 | |
EOMES (Human) Recombinant Protein (Q03) | |
SHQIVPGGRYGVQSFFPEPFVNTLPQARYYNGERTVPQTNGLLSPQQSEEVANPPQRWLVTPVQQPGTNKLDISSYESEYTSSTLLPYGIKSLPLQTSHALGYYPDPTF | |
RUO | |
EOMES | |
Wheat Germ (in vitro) | |
GST | |
Liquid |