missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EPHB3 Partial ORF (NP_004434, 899 a.a. - 997 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002049-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
Sequence: AASLKVIASAQSGMSQPLLDRTVPDYTTFTTVGDWLDAIKMGRYKESFVSAGFASFDLVAQMTAEDLLRIGVTLAGHQKKILSSIQDMRLQMNQTLPVQSpecifications
NP_004434 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
AASLKVIASAQSGMSQPLLDRTVPDYTTFTTVGDWLDAIKMGRYKESFVSAGFASFDLVAQMTAEDLLRIGVTLAGHQKKILSSIQDMRLQMNQTLPVQ | |
RUO | |
EPHB3 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
2049 | |
EPHB3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ETK2|HEK2|TYRO6 | |
EPHB3 | |
Yes |