Learn More
Abnova™ Human EPHB6 Partial ORF (NP_004436, 23 a.a. - 122 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004436 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2051 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16187724
|
Abnova™
H00002051-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16177724
|
Abnova™
H00002051-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. The ephrin receptor encoded by this gene lacks the kinase activity of most receptor tyrosine kinases and binds to ephrin-B ligands. [provided by RefSeq]
Sequence: DTTGETSEIGWLTYPPGGWDEVSVLDDQRRLTRTFEACHVAGAPPGTGQDNWLQTHFVERRGAQRAHIRLHFSVRACSSLGVSGGTCRETFTLYYRQAEESpecifications
NP_004436 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HEP/MGC129910/MGC129911 | |
EPHB6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2051 | |
EPHB6 (Human) Recombinant Protein (Q01) | |
DTTGETSEIGWLTYPPGGWDEVSVLDDQRRLTRTFEACHVAGAPPGTGQDNWLQTHFVERRGAQRAHIRLHFSVRACSSLGVSGGTCRETFTLYYRQAEE | |
RUO | |
EPHB6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |