Learn More
Abnova™ Human EPS8L1 Partial ORF (NP_573441, 181 a.a. - 279 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054869-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is related to epidermal growth factor receptor pathway substrate 8 (EPS8), a substrate for the epidermal growth factor receptor. The function of this protein is unknown. At least two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: RSPAAETPPLQRRPSVRAVISTVERGAGRGRPQAKPIPEAEEAQRPEPVGTSSNADSASPDLGPRGPDLAVLQAEREVDILNHVFDDVESFVSRLQKSASpecifications
NP_573441 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RSPAAETPPLQRRPSVRAVISTVERGAGRGRPQAKPIPEAEEAQRPEPVGTSSNADSASPDLGPRGPDLAVLQAEREVDILNHVFDDVESFVSRLQKSA | |
RUO | |
EPS8L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54869 | |
EPS8L1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DRC3/EPS8R1/FLJ20258/MGC23164/MGC4642/PP10566 | |
EPS8L1 | |
Recombinant | |
wheat germ expression system |