missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Human ErbB2 (HER-2) Fc Chimera Recombinant Protein

Produktkod. 17946331
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
100 μg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 17946331

Brand: Invitrogen™ RP88054

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (1695 μL) be added to the vial to prepare a stock solution of 0.83 mg/mL. Concentration is measured by UV.

ErbB2 (HER2) is a receptor tyrosine kinase that is overexpressed in some breast tumors. Herceptin, used in treatment of metastatic Her2-positive cancer, is monoclonal antibody targeting this kinase.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer P04626
Koncentration 0.25 mg/mL
För användning med (applikation) Control, Binding Assay, Bioactivity
Formatera Lyophilized
Formulering PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gen-ID (Entrez) 2064
Molekylvikt (g/mol) 96.09 kDa
Namn Human ErbB2 (HER-2) Fc Chimera
pH-intervall 7.4
Reningsmetod Purified
Kvantitet 100 μg
Förvaringskrav -20°C, Avoid Freeze/Thaw Cycles
Regulatorisk status RUO
Endotoxinkoncentration <1.0 EU/μg
Gene Alias avian erythroblastosis oncogene B 2; Avian erythroblastosis viral (v-erb-B2) oncogene homologue 2 (neuro/glioblastoma derived oncogene homolog); CD antigen CD340; CD340; c-erb B2/neu protein; c-erbB2; C-erbB-2; c-neu; epidermal growth factor receptor-related protein; erbb 2; ERBB2; erb-b2; Erbb-2; ErbB2 (pTyr1139); ErbB2 (pY1139); ErbB2 phospho Y1139; erb-b2 receptor tyrosine kinase 2; her 2; HER2; HER-2; HER-2/neu; herstatin; human epidermal growth factor receptor 2; Kiaa3023; metas; Metastatic lymph node gene 19 protein; mKIAA3023; MLN 19; MLN19; NEU; Neu oncogene; NEU proto-oncogene; neuro/glioblastoma derived oncogene homolog; neuroblastoma/glioblastoma derived oncogene homolog; NGL; p185erbB2; p185neu; phospho ErbB2; proto-oncogene c-ErbB-2; Proto-oncogene Neu; receptor tyrosine-protein kinase erbB-2; TKR1; Tyrosine kinase-type cell surface receptor HER2; v-erb-b2 avian erythroblastic leukemia viral oncogene 2; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2; v-erb-b2 avian erythroblastic leukemia viral oncoprotein 2; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog; v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastoma derived oncogene homolog (avian)
Vanligt namn Her-2 (ErbB2)
Gensymbol ERBB2
Biologisk aktivitet 1. Immobilized Her2/ERBB2 Protein, Human, Recombinant (ECD, hFc Tag) (at 2 μg/ml (100 μL/well) can bind Anti-Erbb2 Antibody (Trastuzumab), the EC50 is 16-80 ng/mL 2. Measured by its ability to block anti-ErbB2 mediated inhibition of BT474 human breast ductal carcinoma cell proliferation. The ED50 for this effect is 0.5-3 μg/mL in the presence of 0.6 μg/mL Anti-ErbB2/Her2 Monoclonal Antibody.)
Produkttyp Protein
Konjugera Unconjugated
Rekombinant Recombinant
Protein Tag Fc-tag
Sekvens Human ErbB2, amino acids Met1-Thr652 (Accession # NP_004439.2) with a C-terminal human IgG1 Fc region
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Aktivitet Immobilized Recombinant Human HER2 / ErbB2 Protein (Fc Tag) (Cat: 10004-H02H) at 0.05 μg/mL (100 μL/well) can bind Trastuzumab derived Anti-Erbb2 antibody, Human IgG1, the EC50 is 10.2-30.8 ng/mL (QC tested).
Uttryckssystem HEK293 cells
Form Lyophilized
Renhet eller kvalitetsklass ≥90% as determined by SDS-PAGE. ≥90% as determined by SEC-HPLC.
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.