missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Human ERGIC1 (aa 52-125) Control Fragment Recombinant Protein

Produktkod. 30210867
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30210867 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30210867 Leverantör Invitrogen™ Leverantörsnummer RP92410

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53837 (PA5-53837. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The function of ERGIC1 remains largely unknown. ERGIC1 was highly expressed in most primary prostate tumors. It is an intriguing potential drug target especially for the ERG oncogene expressing tumors.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer Q969X5
Koncentration ≥5.0 mg/mL
För användning med (applikation) Blocking Assay, Control
Formulering 1 M urea, PBS with no preservative; pH 7.4
Gen-ID (Entrez) 57222
Namn Human ERGIC1 (aa 52-125) Control Fragment
Kvantitet 100 μL
Regulatorisk status RUO
Gene Alias 1200007D18Rik; endoplasmic reticulum-golgi intermediate compartment (ERGIC) 1; endoplasmic reticulum-golgi intermediate compartment 1; endoplasmic reticulum-golgi intermediate compartment 32 kDa protein; endoplasmic reticulum-Golgi intermediate compartment protein 1; ERGIC1; ERGIC32; ERGIC-32; ER-Golgi intermediate compartment 32 kDa protein; HT034; KIAA1181; maa-136; mKIAA1181 protein-like; NET24
Vanligt namn ERGIC1
Gensymbol ERGIC1
Konjugera Unconjugated
Art Human
Rekombinant Recombinant
Protein Tag His-ABP-tag
Sekvens VVNELYVDDPDKDSGGKIDVSLNISLPNLHCELVGLDIQDEMGRHEVGHIDNSMKIPLNNGAGCRFEGQFSINK
Innehåll och lagring -20°C, Avoid Freeze/Thaw Cycles
Uttryckssystem E. coli
Form Liquid
Renhet eller kvalitetsklass >80% by SDS-PAGE and Coomassie blue staining
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.