missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ERGIC2 Full-length ORF (AAH64522.1, 1 a.a. - 215 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051290-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ERGIC2, or PTX1, is a ubiquitously expressed nuclear protein that is downregulated in prostate carcinoma (Kwok et al., 2001 [PubMed 11445006]).[supplied by OMIM]
Sequence: MRRLNRKKTLSLVKELDAFPKVPESYVETSASGGTVSLIAFTTMALLTIMEFSVYQDTWMKYEYEVDKDFSSKLRINIDITVAMKCQYVGADVLDLAETMFASADGLVYEPTVFDLSPQQKEWQRMLQLIQSRLQEEHSLQDVIFKSAFKSTSTALPPREDDSSQSPNACRIHGHLYVNKVAGNFHITVGQFHILVVMHIWQHLSTMNLTIFLIESpecifications
AAH64522.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDA14/Erv41/MGC111152/PTX1/cd002 | |
ERGIC2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51290 | |
ERGIC2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRRLNRKKTLSLVKELDAFPKVPESYVETSASGGTVSLIAFTTMALLTIMEFSVYQDTWMKYEYEVDKDFSSKLRINIDITVAMKCQYVGADVLDLAETMFASADGLVYEPTVFDLSPQQKEWQRMLQLIQSRLQEEHSLQDVIFKSAFKSTSTALPPREDDSSQSPNACRIHGHLYVNKVAGNFHITVGQFHILVVMHIWQHLSTMNLTIFLIE | |
RUO | |
ERGIC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |