missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human Erythropoietin R Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF-322-PB
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Erythropoietin R Polyclonal specifically detects Erythropoietin R in Human samples. It is validated for Western Blot, Simple Western.
Specifikationer
| Erythropoietin R | |
| Polyclonal | |
| Unconjugated | |
| P19235 | |
| EPOR | |
| Mouse myeloma cell line NS0-derived recombinant human Erythropoietin R Pro26-Pro250 Accession # P19235 | |
| 100 μg | |
| Primary | |
| Detects human Erythropoietin R in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with rhEpo and rhTpo is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| EpoR, EPO-R, erythropoietin receptor, MGC138358 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2057 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering