missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ETS2 Full-length ORF (NP_005230.1, 1 a.a. - 469 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_005230.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2114 |
Molecular Weight (g/mol) | 79.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16177567
|
Abnova™
H00002114-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16167567
|
Abnova™
H00002114-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes (Dwyer et al., 2007 [PubMed 17986575]).[supplied by OMIM]
Sequence: MNDFGIKNMDQVAPVANSYRGTLKRQPAFDTFDGSLFAVFPSLNEEQTLQEVPTGLDSISHDSANCELPLLTPCSKAVMSQALKATFSGFKKEQRRLGIPKNPWLWSEQQVCQWLLWATNEFSLVNVNLQRFGMNGQMLCNLGKERFLELAPDFVGDILWEHLEQMIKENQEKTEDQYEENSHLTSVPHWINSNTLGFGTEQAPYGMQTQNYPKGGLLDSMCPASTPSVLSSEQEFQMFPKSRLSSVSVTYCSVSQDFPGSNLNLLTNNSGTPKDHDSPENGADSFESSDSLLQSWNSQSSLLDVQRVPSFESFEDDCSQSLCLNKPTMSFKDYIQERSDPVEQGKPVIPAAVLAGFTGSGPIQLWQFLLELLSDKSCQSFISWTGDGWEFKLADPDEVARRWGKRKNKPKMNYEKLSRGLRYYYDKNIIHKTSGKRYVYRFVCDLQNLLGFTPEELHAILGVQPDTEDSpecifications
NP_005230.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNDFGIKNMDQVAPVANSYRGTLKRQPAFDTFDGSLFAVFPSLNEEQTLQEVPTGLDSISHDSANCELPLLTPCSKAVMSQALKATFSGFKKEQRRLGIPKNPWLWSEQQVCQWLLWATNEFSLVNVNLQRFGMNGQMLCNLGKERFLELAPDFVGDILWEHLEQMIKENQEKTEDQYEENSHLTSVPHWINSNTLGFGTEQAPYGMQTQNYPKGGLLDSMCPASTPSVLSSEQEFQMFPKSRLSSVSVTYCSVSQDFPGSNLNLLTNNSGTPKDHDSPENGADSFESSDSLLQSWNSQSSLLDVQRVPSFESFEDDCSQSLCLNKPTMSFKDYIQERSDPVEQGKPVIPAAVLAGFTGSGPIQLWQFLLELLSDKSCQSFISWTGDGWEFKLADPDEVARRWGKRKNKPKMNYEKLSRGLRYYYDKNIIHKTSGKRYVYRFVCDLQNLLGFTPEELHAILGVQPDTED | |
RUO | |
ETS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
2114 | |
ETS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ETS2IT1 | |
ETS2 | |
Recombinant | |
wheat germ expression system |