missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EXOC5 Full-length ORF (NP_006535.1, 1 a.a. - 708 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010640-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135482

  • 4515.00 SEK / 10µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity. [provided by RefSeq]

Sequence: MATTAELFEEPFVADEYIERLVWRTPGGGSRGGPEAFDPKRLLEEFVNHIQELQIMDERIQRKVEKLEQQCQKEAKEFAKKVQELQKSNQVAFQHFQELDEHISYVATKVCHLGDQLEGVNTPRQRAVEAQKLMKYFNEFLDGELKSDVFTNSEKIKEAADIIQKLHLIAQELPFDRFSEVKSKIASKYHDLECQLIQEFTSAQRRGEISRMREVAAVLLHFKGYSHCVDVYIKQCQEGAYLRNDIFEDAGILCQRVNKQVGDIFSNPETVLAKLIQNVFEIKLQSFVKEQLEECRKSDAEQYLKNLYDLYTRTTNLSSKLMEFNLGTDKQTFLSKLIKSIFISYLENYIEVETGYLKSRSAMILQRYYDSKNHQKRSIGTGGIQDLKERIRQRTNLPLGPSIDTHGETFLSQEVVVNLLQETKQAFERCHRLSDPSDLPRNAFRIFTILVEFLCIEHIDYALETGLAGIPSSDSRNANLYFLDVVQQANTIFHLFDKQFNDHLMPLISSSPKLSECLQKKKEIIEQMEMKLDTGIDRTLNCMIGQMKHILAAEQKKTDFKPEDENNVLIQYTNACVKVCAYVRKQVEKIKNSMDGKNVDTVLMELGVRFHRLIYEHLQQYSYSCMGGMLAICDVAEYRKCAKDFKIPMVLHLFDTLHALCNLLVVAPDNLKQVCSGEQLANLDKNILHSFVQLRADYRSARLARHFS
Specifications

Specifications

NP_006535.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
108.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp666H126/HSEC10/PRO1912/SEC10/SEC10L1/SEC10P
EXOC5
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10640
EXOC5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATTAELFEEPFVADEYIERLVWRTPGGGSRGGPEAFDPKRLLEEFVNHIQELQIMDERIQRKVEKLEQQCQKEAKEFAKKVQELQKSNQVAFQHFQELDEHISYVATKVCHLGDQLEGVNTPRQRAVEAQKLMKYFNEFLDGELKSDVFTNSEKIKEAADIIQKLHLIAQELPFDRFSEVKSKIASKYHDLECQLIQEFTSAQRRGEISRMREVAAVLLHFKGYSHCVDVYIKQCQEGAYLRNDIFEDAGILCQRVNKQVGDIFSNPETVLAKLIQNVFEIKLQSFVKEQLEECRKSDAEQYLKNLYDLYTRTTNLSSKLMEFNLGTDKQTFLSKLIKSIFISYLENYIEVETGYLKSRSAMILQRYYDSKNHQKRSIGTGGIQDLKERIRQRTNLPLGPSIDTHGETFLSQEVVVNLLQETKQAFERCHRLSDPSDLPRNAFRIFTILVEFLCIEHIDYALETGLAGIPSSDSRNANLYFLDVVQQANTIFHLFDKQFNDHLMPLISSSPKLSECLQKKKEIIEQMEMKLDTGIDRTLNCMIGQMKHILAAEQKKTDFKPEDENNVLIQYTNACVKVCAYVRKQVEKIKNSMDGKNVDTVLMELGVRFHRLIYEHLQQYSYSCMGGMLAICDVAEYRKCAKDFKIPMVLHLFDTLHALCNLLVVAPDNLKQVCSGEQLANLDKNILHSFVQLRADYRSARLARHFS
RUO
EXOC5
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers