missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EXOC7 Partial ORF (NP_001013861, 586 a.a. - 684 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_001013861 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23265 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16174526
|
Abnova™
H00023265-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16184526
|
Abnova™
H00023265-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
EXOC7 is a component of the exocyst, which is an evolutionarily conserved octameric protein complex essential for exocytosis. The exocyst targets secretory vesicles at specific domains of the plasma membrane for cell surface expansion and protein secretion (Zuo et al., 2006 [PubMed 17086175]).[supplied by OMIM]
Sequence: VFQPGVKLRDKERQIIKERFKGFNDGLEELCKIQKAWAIPDTEQRDRIRQAQKTIVKETYGAFLQKFGSVPFTKNPEKYIKYGVEQVGDMIDRLFDTSASpecifications
NP_001013861 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
2-5-3p/DKFZp686J04253/EX070/EXO70/EXOC1/Exo70p/FLJ40965/FLJ46415/YJL085W | |
EXOC7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23265 | |
EXOC7 (Human) Recombinant Protein (Q01) | |
VFQPGVKLRDKERQIIKERFKGFNDGLEELCKIQKAWAIPDTEQRDRIRQAQKTIVKETYGAFLQKFGSVPFTKNPEKYIKYGVEQVGDMIDRLFDTSA | |
RUO | |
EXOC7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |