Learn More
Abnova™ Human EYA4 Full-length ORF (NP_742101.2, 1 a.a. - 616 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_742101.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2070 |
Molecular Weight (g/mol) | 93.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16117517
|
Abnova™
H00002070-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16107517
|
Abnova™
H00002070-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the eyes absent (EYA) family of proteins. The encoded protein may act as a transcriptional activator through its protein phosphatase activity, and it may be important for eye development, and for continued function of the mature organ of Corti. Mutations in this gene are associated with postlingual, progressive, autosomal dominant hearing loss at the deafness, autosomal dominant nonsyndromic sensorineural 10 locus. Defects in this gene are also associated with dilated cardiomyopathy 1J. Three transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MEDSQDLNEQSVKKTCTESDVSQSQNSRSMEMQDLASPHTLVGGGDTPGSSKLEKSNLSSTSVTTNGTGVSLLAVKTEPLNSSETTATTGDGALDTFTGSVITSSGYSPRSAHQYSPQLYPSKPYPHILSTPAAQTMSAYAGQTQYSGMQQPAVYTAYSQTGQPYSLPTYDLGVMLPAIKTESGLSQTQSPLQSGCLSYSPGFSTPQPGQTPYSYQMPGSSFAPSSTIYANNSVSNSTNFSGSQQDYPSYTAFGQNQYAQYYSASTYGAYMTSNNTADGTPSSTSTYQLQESLPGLTNQPGEFDTMQSPSTPIKDLDERTCRSSGSKSRGRGRKNNPSPPPDSDLERVFVWDLDETIIVFHSLLTGSYAQKYGKDPPMAVTLGLRMEEMIFNLADTHLFFNDLEECDQVHIDDVSSDDNGQDLSTYSFATDGFHAAASSANLCLPTGVRGGVDWMRKLAFRYRRVKELYNTYKNNVGGLLGPAKRDAWLQLRAEIEGLTDSWLTNALKSLSIISTRSNCINVLVTTTQLIPALAKVLLYSLGGAFPIENIYSATKIGKESCFERIMQRFGRKVVYVVIGDGVEEEQAAKKHNMPFWRISSHSDLLALHQALELEYLSpecifications
NP_742101.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
93.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEDSQDLNEQSVKKTCTESDVSQSQNSRSMEMQDLASPHTLVGGGDTPGSSKLEKSNLSSTSVTTNGTGVSLLAVKTEPLNSSETTATTGDGALDTFTGSVITSSGYSPRSAHQYSPQLYPSKPYPHILSTPAAQTMSAYAGQTQYSGMQQPAVYTAYSQTGQPYSLPTYDLGVMLPAIKTESGLSQTQSPLQSGCLSYSPGFSTPQPGQTPYSYQMPGSSFAPSSTIYANNSVSNSTNFSGSQQDYPSYTAFGQNQYAQYYSASTYGAYMTSNNTADGTPSSTSTYQLQESLPGLTNQPGEFDTMQSPSTPIKDLDERTCRSSGSKSRGRGRKNNPSPPPDSDLERVFVWDLDETIIVFHSLLTGSYAQKYGKDPPMAVTLGLRMEEMIFNLADTHLFFNDLEECDQVHIDDVSSDDNGQDLSTYSFATDGFHAAASSANLCLPTGVRGGVDWMRKLAFRYRRVKELYNTYKNNVGGLLGPAKRDAWLQLRAEIEGLTDSWLTNALKSLSIISTRSNCINVLVTTTQLIPALAKVLLYSLGGAFPIENIYSATKIGKESCFERIMQRFGRKVVYVVIGDGVEEEQAAKKHNMPFWRISSHSDLLALHQALELEYL | |
RUO | |
EYA4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
2070 | |
EYA4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CMD1J/DFNA10 | |
EYA4 | |
Recombinant | |
wheat germ expression system |