missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FBLN2 Full-length ORF (NP_001989.2, 28 a.a. - 1184 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_001989.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2199 |
Molecular Weight (g/mol) | 152.90kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16194861
|
Abnova™
H00002199-P02.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16184861
|
Abnova™
H00002199-P02.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes an extracellular matrix protein, which belongs to the fibulin family. This protein binds various extracellular ligands and calcium. It may play a role during organ development, in particular, during the differentiation of heart, skeletal and neuronal structures. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: APRQDCTGVECPPLENCIEEALEPGACCATCVQQGCACEGYQYYDCLQGGFVRGRVPAGQSYFVDFGSTECSCPPGGGKISCQFMLCPELPPNCIEAVVVADSCPQCGQVGCVHAGHKYAAGHTVHLPPCRACHCPDAGGELICYQLPGCHGNFSDAEEGDPERHYEDPYSYDQEVAEVEAATALGGEVQAGAVQAGAGGPPAALGGGSQPLSTIQAPPWPAVLPRPTAAAALGPPAPVQAKARRVTEDSEEEEEEEEEREEMAVTEQLAAGGHRGLDGLPTTAPAGPSLPIQEERAEAGARAEAGARPEENLILDAQATSRSTGPEGVTHAPSLGKAALVPTQAVPGSPRDPVKPSPHNILSTSLPDAAWIPPTREVPRKPQVLPHSHVEEDTDPNSVHSIPRSSPEGSTKDLIETCCAAGQQWAIDNDECLEIPESGTEDNVCRTAQRHCCVSYLQEKSCMAGVLGAKEGETCGAEDNDSCGISLYKQCCDCCGLGLRVRAEGQSCESNPNLGYPCNHVMLSCCEGEEPLIVPEVRRPPEPAAAPRRVSEAEMAGREALSLGTEAELPNSLPGDDQDECLLLPGELCQHLCINTVGSYHCACFPGFSLQDDGRTCRPEGHPPQPEAPQEPALKSEFSQVASNTIPLPLPQPNTCKDNGPCKQVCSTVGGSAICSCFPGYAIMADGVSCEDINECVTDLHTCSRGEHCVNTLGSFHCYKALTCEPGYALKDGECEDVDECAMGTHTCQPGFLCQNTKGSFYCQARQRCMDGFLQDPEGNCVDINECTSLSEPCRPGFSCINTVGSYTCQRNPLICARGYHASDDGTKCVDVNECETGVHRCGEGQVCHNLPGSYRCDCKAGFQRDAFGRGCIDVNECWASPGRLCQHTCENTLGSYRCSCASGFLLAADGKRCEDVNECEAQRCSQECANIYGSYQCYCRQGYQLAEDGHTCTDIDECAQGAGILCTFRCLNVPGSYQCACPEQGYTMTANGRSCKDVDECALGTHNCSEAETCHNIQGSFRCLRFECPPNYVQVSKTKCERTTCHDFLECQNSPARITHYQLNFQTGLLVPAHIFRIGPAPAFTGDTIALNIIKGNEEGYFGTRRLNAYTGVVYLQRAVLEPRDFALDVEMKLWRQGSVTTFLAKMHIFFTTFALSpecifications
NP_001989.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
152.90kDa | |
Glutathione Sepharose 4 Fast Flow | |
APRQDCTGVECPPLENCIEEALEPGACCATCVQQGCACEGYQYYDCLQGGFVRGRVPAGQSYFVDFGSTECSCPPGGGKISCQFMLCPELPPNCIEAVVVADSCPQCGQVGCVHAGHKYAAGHTVHLPPCRACHCPDAGGELICYQLPGCHGNFSDAEEGDPERHYEDPYSYDQEVAEVEAATALGGEVQAGAVQAGAGGPPAALGGGSQPLSTIQAPPWPAVLPRPTAAAALGPPAPVQAKARRVTEDSEEEEEEEEEREEMAVTEQLAAGGHRGLDGLPTTAPAGPSLPIQEERAEAGARAEAGARPEENLILDAQATSRSTGPEGVTHAPSLGKAALVPTQAVPGSPRDPVKPSPHNILSTSLPDAAWIPPTREVPRKPQVLPHSHVEEDTDPNSVHSIPRSSPEGSTKDLIETCCAAGQQWAIDNDECLEIPESGTEDNVCRTAQRHCCVSYLQEKSCMAGVLGAKEGETCGAEDNDSCGISLYKQCCDCCGLGLRVRAEGQSCESNPNLGYPCNHVMLSCCEGEEPLIVPEVRRPPEPAAAPRRVSEAEMAGREALSLGTEAELPNSLPGDDQDECLLLPGELCQHLCINTVGSYHCACFPGFSLQDDGRTCRPEGHPPQPEAPQEPALKSEFSQVASNTIPLPLPQPNTCKDNGPCKQVCSTVGGSAICSCFPGYAIMADGVSCEDINECVTDLHTCSRGEHCVNTLGSFHCYKALTCEPGYALKDGECEDVDECAMGTHTCQPGFLCQNTKGSFYCQARQRCMDGFLQDPEGNCVDINECTSLSEPCRPGFSCINTVGSYTCQRNPLICARGYHASDDGTKCVDVNECETGVHRCGEGQVCHNLPGSYRCDCKAGFQRDAFGRGCIDVNECWASPGRLCQHTCENTLGSYRCSCASGFLLAADGKRCEDVNECEAQRCSQECANIYGSYQCYCRQGYQLAEDGHTCTDIDECAQGAGILCTFRCLNVPGSYQCACPEQGYTMTANGRSCKDVD | |
RUO | |
FBLN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2199 | |
FBLN2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBLN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |