Learn More
Abnova™ Human FBLN2 Full-length ORF (NP_001989.2, 28 a.a. - 1184 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002199-P02.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes an extracellular matrix protein, which belongs to the fibulin family. This protein binds various extracellular ligands and calcium. It may play a role during organ development, in particular, during the differentiation of heart, skeletal and neuronal structures. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: APRQDCTGVECPPLENCIEEALEPGACCATCVQQGCACEGYQYYDCLQGGFVRGRVPAGQSYFVDFGSTECSCPPGGGKISCQFMLCPELPPNCIEAVVVADSCPQCGQVGCVHAGHKYAAGHTVHLPPCRACHCPDAGGELICYQLPGCHGNFSDAEEGDPERHYEDPYSYDQEVAEVEAATALGGEVQAGAVQAGAGGPPAALGGGSQPLSTIQAPPWPAVLPRPTAAAALGPPAPVQAKARRVTEDSEEEEEEEEEREEMAVTEQLAAGGHRGLDGLPTTAPAGPSLPIQEERAEAGARAEAGARPEENLILDAQATSRSTGPEGVTHAPSLGKAALVPTQAVPGSPRDPVKPSPHNILSTSLPDAAWIPPTREVPRKPQVLPHSHVEEDTDPNSVHSIPRSSPEGSTKDLIETCCAAGQQWAIDNDECLEIPESGTEDNVCRTAQRHCCVSYLQEKSCMAGVLGAKEGETCGAEDNDSCGISLYKQCCDCCGLGLRVRAEGQSCESNPNLGYPCNHVMLSCCEGEEPLIVPEVRRPPEPAAAPRRVSEAEMAGREALSLGTEAELPNSLPGDDQDECLLLPGELCQHLCINTVGSYHCACFPGFSLQDDGRTCRPEGHPPQPEAPQEPALKSEFSQVASNTIPLPLPQPNTCKDNGPCKQVCSTVGGSAICSCFPGYAIMADGVSCEDINECVTDLHTCSRGEHCVNTLGSFHCYKALTCEPGYALKDGECEDVDECAMGTHTCQPGFLCQNTKGSFYCQARQRCMDGFLQDPEGNCVDINECTSLSEPCRPGFSCINTVGSYTCQRNPLICARGYHASDDGTKCVDVNECETGVHRCGEGQVCHNLPGSYRCDCKAGFQRDAFGRGCIDVNECWASPGRLCQHTCENTLGSYRCSCASGFLLAADGKRCEDVNECEAQRCSQECANIYGSYQCYCRQGYQLAEDGHTCTDIDECAQGAGILCTFRCLNVPGSYQCACPEQGYTMTANGRSCKDVDECALGTHNCSEAETCHNIQGSFRCLRFECPPNYVQVSKTKCERTTCHDFLECQNSPARITHYQLNFQTGLLVPAHIFRIGPAPAFTGDTIALNIIKGNEEGYFGTRRLNAYTGVVYLQRAVLEPRDFALDVEMKLWRQGSVTTFLAKMHIFFTTFALSpecifications
NP_001989.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
152.90kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBLN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
2199 | |
FBLN2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
APRQDCTGVECPPLENCIEEALEPGACCATCVQQGCACEGYQYYDCLQGGFVRGRVPAGQSYFVDFGSTECSCPPGGGKISCQFMLCPELPPNCIEAVVVADSCPQCGQVGCVHAGHKYAAGHTVHLPPCRACHCPDAGGELICYQLPGCHGNFSDAEEGDPERHYEDPYSYDQEVAEVEAATALGGEVQAGAVQAGAGGPPAALGGGSQPLSTIQAPPWPAVLPRPTAAAALGPPAPVQAKARRVTEDSEEEEEEEEEREEMAVTEQLAAGGHRGLDGLPTTAPAGPSLPIQEERAEAGARAEAGARPEENLILDAQATSRSTGPEGVTHAPSLGKAALVPTQAVPGSPRDPVKPSPHNILSTSLPDAAWIPPTREVPRKPQVLPHSHVEEDTDPNSVHSIPRSSPEGSTKDLIETCCAAGQQWAIDNDECLEIPESGTEDNVCRTAQRHCCVSYLQEKSCMAGVLGAKEGETCGAEDNDSCGISLYKQCCDCCGLGLRVRAEGQSCESNPNLGYPCNHVMLSCCEGEEPLIVPEVRRPPEPAAAPRRVSEAEMAGREALSLGTEAELPNSLPGDDQDECLLLPGELCQHLCINTVGSYHCACFPGFSLQDDGRTCRPEGHPPQPEAPQEPALKSEFSQVASNTIPLPLPQPNTCKDNGPCKQVCSTVGGSAICSCFPGYAIMADGVSCEDINECVTDLHTCSRGEHCVNTLGSFHCYKALTCEPGYALKDGECEDVDECAMGTHTCQPGFLCQNTKGSFYCQARQRCMDGFLQDPEGNCVDINECTSLSEPCRPGFSCINTVGSYTCQRNPLICARGYHASDDGTKCVDVNECETGVHRCGEGQVCHNLPGSYRCDCKAGFQRDAFGRGCIDVNECWASPGRLCQHTCENTLGSYRCSCASGFLLAADGKRCEDVNECEAQRCSQECANIYGSYQCYCRQGYQLAEDGHTCTDIDECAQGAGILCTFRCLNVPGSYQCACPEQGYTMTANGRSCKDVD | |
RUO | |
FBLN2 | |
Recombinant | |
wheat germ expression system |