Learn More
Abnova™ Human FBLN5 Full-length ORF (BAG52073.1, 1 a.a. - 448 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010516-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a secreted, extracellular matrix protein containing an Arg-Gly-Asp (RGD) motif and calcium-binding EGF-like domains. It promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. It is prominently expressed in developing arteries but less so in adult vessels. However, its expression is reinduced in balloon-injured vessels and atherosclerotic lesions, notably in intimal vascular smooth muscle cells and endothelial cells. Therefore, the protein encoded by this gene may play a role in vascular development and remodeling. Defects in this gene are a cause of autosomal dominant cutis laxa, autosomal recessive cutis laxa type I (CL type I), and age-related macular degeneration type 3 (ARMD3). [provided by RefSeq]
Sequence: MPGIKRILTVTILALCLPSPGNAQAQCTNGFDLDRQSGQCLDIDECRTIPEACRGDMMCVNQNGGYLCIPRTNPVYRGPYSNPYSTPYSGPYPAAAPPLSAPNYPTISRPLICRFGYQMDESNQCVDVDECATDSHQCNPTQICINTEGGYTCSCTDGYWLLEGQCLDIDECRYGYCQQLCANVPGSYSCTCNPGFTLNEDGRSCQDVNECATENPCVQTCVNTYGSFICRCDPGYELEEDGVHCSDMDECSFSEFLCQHECVNQPGTYFCSCPPGYILLDDNRSCQDINECEHRNHTCNLQQTCYNLQGGFKCIDPIRCEEPYLRISDNRCMCPAENPGCRDQPFTILYRDMDVVSGRSVPADIFQMQATTRYPGAYYIFQIKSGNEGREFYMRQTGPISATLVMTRPIKGPREIQLDLEMITVNTVINFRGSSVIRLRIYVSQYPFSpecifications
BAG52073.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARMD3/DANCE/EVEC/FIBL-5/FLJ90059/UP50 | |
FBLN5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10516 | |
FBLN5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MPGIKRILTVTILALCLPSPGNAQAQCTNGFDLDRQSGQCLDIDECRTIPEACRGDMMCVNQNGGYLCIPRTNPVYRGPYSNPYSTPYSGPYPAAAPPLSAPNYPTISRPLICRFGYQMDESNQCVDVDECATDSHQCNPTQICINTEGGYTCSCTDGYWLLEGQCLDIDECRYGYCQQLCANVPGSYSCTCNPGFTLNEDGRSCQDVNECATENPCVQTCVNTYGSFICRCDPGYELEEDGVHCSDMDECSFSEFLCQHECVNQPGTYFCSCPPGYILLDDNRSCQDINECEHRNHTCNLQQTCYNLQGGFKCIDPIRCEEPYLRISDNRCMCPAENPGCRDQPFTILYRDMDVVSGRSVPADIFQMQATTRYPGAYYIFQIKSGNEGREFYMRQTGPISATLVMTRPIKGPREIQLDLEMITVNTVINFRGSSVIRLRIYVSQYPF | |
RUO | |
FBLN5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |