Learn More
Abnova™ Human FBLN5 Full-length ORF (BAG52073.1, 1 a.a. - 448 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | BAG52073.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10516 |
Molecular Weight (g/mol) | 76.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16132686
|
Abnova™
H00010516-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16142686
|
Abnova™
H00010516-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a secreted, extracellular matrix protein containing an Arg-Gly-Asp (RGD) motif and calcium-binding EGF-like domains. It promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. It is prominently expressed in developing arteries but less so in adult vessels. However, its expression is reinduced in balloon-injured vessels and atherosclerotic lesions, notably in intimal vascular smooth muscle cells and endothelial cells. Therefore, the protein encoded by this gene may play a role in vascular development and remodeling. Defects in this gene are a cause of autosomal dominant cutis laxa, autosomal recessive cutis laxa type I (CL type I), and age-related macular degeneration type 3 (ARMD3). [provided by RefSeq]
Sequence: MPGIKRILTVTILALCLPSPGNAQAQCTNGFDLDRQSGQCLDIDECRTIPEACRGDMMCVNQNGGYLCIPRTNPVYRGPYSNPYSTPYSGPYPAAAPPLSAPNYPTISRPLICRFGYQMDESNQCVDVDECATDSHQCNPTQICINTEGGYTCSCTDGYWLLEGQCLDIDECRYGYCQQLCANVPGSYSCTCNPGFTLNEDGRSCQDVNECATENPCVQTCVNTYGSFICRCDPGYELEEDGVHCSDMDECSFSEFLCQHECVNQPGTYFCSCPPGYILLDDNRSCQDINECEHRNHTCNLQQTCYNLQGGFKCIDPIRCEEPYLRISDNRCMCPAENPGCRDQPFTILYRDMDVVSGRSVPADIFQMQATTRYPGAYYIFQIKSGNEGREFYMRQTGPISATLVMTRPIKGPREIQLDLEMITVNTVINFRGSSVIRLRIYVSQYPFSpecifications
BAG52073.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPGIKRILTVTILALCLPSPGNAQAQCTNGFDLDRQSGQCLDIDECRTIPEACRGDMMCVNQNGGYLCIPRTNPVYRGPYSNPYSTPYSGPYPAAAPPLSAPNYPTISRPLICRFGYQMDESNQCVDVDECATDSHQCNPTQICINTEGGYTCSCTDGYWLLEGQCLDIDECRYGYCQQLCANVPGSYSCTCNPGFTLNEDGRSCQDVNECATENPCVQTCVNTYGSFICRCDPGYELEEDGVHCSDMDECSFSEFLCQHECVNQPGTYFCSCPPGYILLDDNRSCQDINECEHRNHTCNLQQTCYNLQGGFKCIDPIRCEEPYLRISDNRCMCPAENPGCRDQPFTILYRDMDVVSGRSVPADIFQMQATTRYPGAYYIFQIKSGNEGREFYMRQTGPISATLVMTRPIKGPREIQLDLEMITVNTVINFRGSSVIRLRIYVSQYPF | |
RUO | |
FBLN5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10516 | |
FBLN5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARMD3/DANCE/EVEC/FIBL-5/FLJ90059/UP50 | |
FBLN5 | |
Recombinant | |
wheat germ expression system |