Learn More
Abnova™ Human FBXL21 Full-length ORF (AAH44938.1, 1 a.a. - 70 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH44938.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26223 |
Molecular Weight (g/mol) | 34.1kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16188172
|
Abnova™
H00026223-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16178172
|
Abnova™
H00026223-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains 6 tandem leucine-rich repeats. The amino acid sequence of this protein is highly similar to that of f-box and leucine-rich repeat protein 3A. Comparisons of this gene to orthologous sequences suggest that it may be a pseudogene, and may no longer express a functional protein.[provided by RefSeq]
Sequence: MNVSESHFVSALTVFINSKSLSSIKIEDTPVDDPSLKILVANNSDTLRLLKMSSCPHVSSDDGLHFLKLESpecifications
AAH44938.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNVSESHFVSALTVFINSKSLSSIKIEDTPVDDPSLKILVANNSDTLRLLKMSSCPHVSSDDGLHFLKLE | |
RUO | |
FBXL21 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
26223 | |
FBXL21 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBL3B/FBXL3B/FBXL3P/Fbl21/MGC120237 | |
FBXL21 | |
Recombinant | |
wheat germ expression system |