Learn More
Abnova™ Human FBXO44 Partial ORF (NP_001014765, 168 a.a. - 255 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00093611-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It is also a member of the NFB42 (neural F Box 42 kDa) family, similar to F-box only protein 2 and F-box only protein 6. Four alternatively spliced transcript variants encoding two distinct isoforms have been found for this gene. [provided by RefSeq]
Sequence: PDCGSKYQLCVQLLSSAHAPLGTFQPDPATIQQKSDAKWREVSHTFSNYPPGVRYIWFQHGGVDTHYWAGWYGPRVTNSSITIGPPLPSpecifications
NP_001014765 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PDCGSKYQLCVQLLSSAHAPLGTFQPDPATIQQKSDAKWREVSHTFSNYPPGVRYIWFQHGGVDTHYWAGWYGPRVTNSSITIGPPLP | |
RUO | |
FBXO44 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
93611 | |
FBXO44 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781J0852/FBG3/FBX30/FBX6A/Fbx44/Fbxo6a/MGC14140 | |
FBXO44 | |
Recombinant | |
wheat germ expression system |