missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXW5 Full-length ORF (AAH00850.1, 1 a.a. - 159 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054461-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16130543

  • 3343.96 SEK / 10µg

Estimated Shipment: 09-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene contains WD-40 domains, in addition to an F-box motif, so it belongs to the Fbw class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene, however, they were found to be nonsense-mediated mRNA decay (NMD) candidates, hence not represented. [provided by RefSeq]

Sequence: MGLSPDNRYLYVNSRAWPNGAVVADPMQPPPIAEEIDLLVFDLKTMREVRRALRAHRAYTPNDECFFIFLDVSRDFVASGAEDRHGYIWDRHYNICLARLRHEDVVNSVVFSPQEQELLLTASDDATIKAWRSPRTMRVLQAPRPRPRTFFSWLASQRR
Specifications

Specifications

AAH00850.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434B205/Fbw5/MGC20962/RP11-229P13.10
FBXW5
Yes
wheat germ expression system
Liquid
54461
FBXW5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGLSPDNRYLYVNSRAWPNGAVVADPMQPPPIAEEIDLLVFDLKTMREVRRALRAHRAYTPNDECFFIFLDVSRDFVASGAEDRHGYIWDRHYNICLARLRHEDVVNSVVFSPQEQELLLTASDDATIKAWRSPRTMRVLQAPRPRPRTFFSWLASQRR
RUO
FBXW5
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates