missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fc Receptor Binding Inhibitor anti-Human, Functional Grade, Polyclonal, eBioscience™

Polyclonal Antibody

Brand:  Invitrogen™ 16-9161-71

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15218827

  • 926.00 SEK / 25 tests


Estimated Shipment
03-11-2020
Add to basket

Description

Description

Use in sterile environment
Specifications

Specifications

Flow Cytometric Analysis
25 tests
Human
Aqueous buffer, no sodium azide
2° to 8°C
Videos
Documents

For Research Use Only