Learn More
Abnova™ Human FCRL1 Full-length ORF (NP_443170.1, 1 a.a. - 429 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00115350-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains three extracellular C2-like immunoglobulin domains, a transmembrane domain and a cytoplasmic domain with two immunoreceptor-tyrosine activation motifs. This protein may play a role in the regulation of cancer cell growth. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MLPRLLLLICAPLCEPAELFLIASPSHPTEGSPVTLTCKMPFLQSSDAQFQFCFFRDTRALGPGWSSSPKLQIAAMWKEDTGSYWCEAQTMASKVLRSRRSQINVHRVPVADVSLETQPPGGQVMEGDRLVLICSVAMGTGDITFLWYKGAVGLNLQSKTQRSLTAEYEIPSVRESDAEQYYCVAENGYGPSPSGLVSITVRIPVSRPILMLRAPRAQAAVEDVLELHCEALRGSPPILYWFYHEDITLGSRSAPSGGGASFNLSLTEEHSGNYSCEANNGLGAQRSEAVTLNFTVPTGARSNHLTSGVIEGLLSTLGPATVALLFCYGLKRKIGRRSARDPLRSLPSPLPQEFTYLNSPTPGQLQPIYENVNVVSGDEVYSLAYYNQPEQESVAAETLGTHMEDKVSLDIYSRLRKANITDVDYEDAMSpecifications
NP_443170.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp667O1421/FCRH1/IFGP1/IRTA5/RP11-367J7.7 | |
FCRL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
115350 | |
FCRL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLPRLLLLICAPLCEPAELFLIASPSHPTEGSPVTLTCKMPFLQSSDAQFQFCFFRDTRALGPGWSSSPKLQIAAMWKEDTGSYWCEAQTMASKVLRSRRSQINVHRVPVADVSLETQPPGGQVMEGDRLVLICSVAMGTGDITFLWYKGAVGLNLQSKTQRSLTAEYEIPSVRESDAEQYYCVAENGYGPSPSGLVSITVRIPVSRPILMLRAPRAQAAVEDVLELHCEALRGSPPILYWFYHEDITLGSRSAPSGGGASFNLSLTEEHSGNYSCEANNGLGAQRSEAVTLNFTVPTGARSNHLTSGVIEGLLSTLGPATVALLFCYGLKRKIGRRSARDPLRSLPSPLPQEFTYLNSPTPGQLQPIYENVNVVSGDEVYSLAYYNQPEQESVAAETLGTHMEDKVSLDIYSRLRKANITDVDYEDAM | |
RUO | |
FCRL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |