Learn More
Abnova™ Human FER1L3 Partial ORF (NP_038479, 655 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_038479 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26509 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16185516
|
Abnova™
H00026509-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16175516
|
Abnova™
H00026509-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mutations in dysferlin, a protein associated with the plasma membrane, can cause muscle weakness that affects both proximal and distal muscles. The protein encoded by this gene is a type II membrane protein that is structurally similar to dysferlin. It is a member of the ferlin family and associates with both plasma and nuclear membranes. The protein contains C2 domains that play a role in calcium-mediated membrane fusion events, suggesting that it may be involved in membrane regeneration and repair. Two transcript variants encoding different isoforms have been found for this gene. Other possible variants have been detected, but their full-length nature has not been determined. [provided by RefSeq]
Sequence: DAVNTLLAMAERLQTNIEALKSGIQGKIPANQLAELWLKLIDEVIEDTRYTLPLTEGKANVTVLDTQIRKLRSRSLSQIHEAAVRMRSEATDVKSTLAEISpecifications
NP_038479 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FER1L3/FLJ36571/FLJ90777 | |
MYOF | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
26509 | |
FER1L3 (Human) Recombinant Protein (Q01) | |
DAVNTLLAMAERLQTNIEALKSGIQGKIPANQLAELWLKLIDEVIEDTRYTLPLTEGKANVTVLDTQIRKLRSRSLSQIHEAAVRMRSEATDVKSTLAEI | |
RUO | |
MYOF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |