missing translation for 'onlineSavingsMsg'
Läs mer

Human FGFR5/FGFRL1 Antibody, R&D Systems™
GREENER_CHOICE

Produktkod. 18787207
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18787207 25 μg
18062632 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18787207 Leverantör R&D Systems Leverantörsnummer AF2257SP

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Goat Polyclonal Antibody

FGFR5/FGFRL1 Polyclonal specifically detects FGFR5/FGFRL1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen FGFR5/FGFRL1
Användningsområden Western Blot
Klassificering Polyclonal
Koncentration LYOPH
Konjugera Unconjugated
Utspädning Western Blot 0.1 ug/mL
Formulering Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Genaccessionsnr. Q8N441
Gene Alias FGF homologous factor receptor, FGF R5, FGF receptor-like protein 1, FGFR5, FGFR-5, FGFRL1, FGFR-like protein, FHFR, Fibroblast growth factor receptor 5, fibroblast growth factor receptor-like 1
Gensymboler FGFRL1
Värd art Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human FGF R5/FGFRL1 Ala25-Pro378 Accession # Q8N441
Reningsmetod Affinity Purified
Kvantitet 25 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 53834
Testspecificitet Detects human FGF R5/FGFRL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 65% cross-reactivity with recombinant mouse (rm) FGF R5 and rmFGF R5 beta is observed and less than 1% cross-reactivity with recombinant human (rh) FGF R1 alpha (IIIb), rhFGF R1 alpha (IIIc), rhFGF R1 beta (IIIb), rhFGF R1 beta (IIIc), rhFGF R2 alpha (IIIb), rhFGF R2 alpha (IIIc), rhFGF R2 beta (IIIb), rhFGF R2 beta (IIIc), rhFGF R3 alpha (IIIb), rhFGF R3 (IIIc), and rhFGF R4 is observed.
Rekonstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Målarter Human
Innehåll och lagring Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotyp IgG
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.